Oakton® DO 6+ Dissolved Oxygen Meters
High accuracy combined with economical pricing
  • Low-maintenance, galvanic probe requires no warm-up time
  • Includes protective rubber boot which also functions as a benchtop stand
This meter is a great value. The ranges work to test water quality in wastewater, surface water, groundwater, or drinking water. LCD shows results in ppm or mg/L with enunciators to aid in use and troubleshooting. Six simple buttons allow for freezing measurements, toggling between display units and temperature results, setup, calibration, and automatic barometric and salinity compensation after manual parameter inputs. Additional benefits of the simple design allow you to view electrode slope, zero offset, and millivolt values for electrode diagnostics.
Purchase zero oxygen solution (00653-00) separately to calibrate meter.
ITEM(S) ON SALE
$535.00 - $986.50USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Accessories Included NIST Calibration Included Probe Included
Availability Pricing
COMPARE
No Yes No
$535.00USD / Each
COMPARE
No No Yes
$794.75USD / Each
$935.00USD / Each
COMPARE
No Yes Yes
$893.50USD / Each
COMPARE
Yes No Yes
$892.50USD / Each
$1,050.00USD / Each
COMPARE
Yes Yes Yes
$986.50USD / Each