Oakton® Benchtop CON 2700 Meter
Advanced meter for benchtop conductivity measurement!
  • Space-saving design takes up less room on the bench than other meters
  • Large display lets you see your measurements from across the lab
More features of the Oakton Benchtop CON 2700 MeterOakton CON 2700 Meter features adjustable temperature coefficient for better accuracy, and selectable cell constant for very high or low range measurements. Selectable manual or automatic temperature compensation gives greater precision. Water-resistant membrane keypad and convenient pull-out reference guide provide useful features for the lab.
Meter has advanced features for GLP-compliant readings—all calibration and stored data are stamped with time/date. Advanced setup options allow you to customize the meter to your needs. Informative display shows measurements together with temperature, electrode status, calibration points, time, and date all at once! Bright backlight helps you view readings under all conditions. Visual stability indicator informs you when measurements have stabilized, eliminating guesswork. Nonvolatile memory stores up to 500 data sets. Programmable functions include datalogging intervals, limit alarms, and password protection. Data is easily exported to a computer via RS-232 port. Order RS-232 cable and optional RS-232 to USB adapter separately.
For full specifications and operating instructions please view the instruction manual.
ITEM(S) ON SALE
$951.57 - $1,261.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Benchtop Meter with Probe and Stand
$951.57USD / Each
$1,119.50USD / Each
COMPARE
meter w/probe & stand with NIST-traceable calibvration
$1,261.50USD / Each