Oakton® 550 Series Benchtop Meter Accessories
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
$87.00 - $276.75USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Replacement pH Electrode with Temperature Sensor
$87.00USD / Each
COMPARE
Replacement Conductivity Cell K= 1.0
$140.00USD / Each
COMPARE
Replacement Conductivity Cell K= 10.0
$150.75USD / Each
COMPARE
Replacement Conductivity Cell K= 0.1
$276.75USD / Each