Nova-Strobe Deluxe Digital Stroboscopes
Phase shifting models let you use your stroboscopes continuously
  • View different parts of moving machinery
These stroboscopes feature a pistol-grip handle with a latching trigger and a 1/4–20 threaded bushing for extended “hands-free” tripod mounting. High flash energy allows measurements in bright areas. Select standard or phase—shifting models according to your application.
$673.00 - $1,155.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Display Type Flash Duration
Availability Pricing
Monarch 6203-010 Nova-Strobe DAX Deluxe Digital Stroboscope; 115 VAC EW-87701-15 Mfr # 6203-010
COMPARE
6-digit numeric, 5-digit alphanumeric backlit LCD 8 to 20 µsec
$673.00USD / Each
Monarch 6203-011 Nova Strobe dax Stroboscope Kit, 115 VAC, with spare lamp and carrying case EW-87701-16 Mfr # 6203-011
COMPARE
6-digit numeric, 5-digit alphanumeric backlit LCD 8 to 20 µsec
$863.00USD / Each
Monarch 6203-012 Nova-Strobe DAX Deluxe Digital Stroboscope; 230 VAC EW-87701-17 Mfr # 6203-012
COMPARE
6-digit numeric, 5-digit alphanumeric backlit LCD 8 to 20 µsec
$735.00USD / Each
Monarch 6204-012 Nova Strobe dbx Deluxe Stroboscope, with 115/230 VAC recharger EW-87701-21 Mfr # 6204-012
COMPARE
6-digit numeric, 5-digit alphanumeric backlit LCD 8 to 20 µsec
$753.00USD / Each
Monarch 6204-013 Nova-Strobe DBX Deluxe Stroboscope Kit EW-87701-22 Mfr # 6204-013
COMPARE
6-digit numeric, 5-digit alphanumeric backlit LCD 8 to 20 µsec
$910.00USD / Each
Monarch 6210-020 Nova-Strobe PBX Phase-Shifting Stroboscope EW-87701-23 Mfr # 6210-020
COMPARE
6-digit numeric, 5-digit alphanumeric backlit LCD 8 to 20 µsec
$955.50USD / Each
Monarch 6210-021 Nova-Strobe PBX Deluxe Stroboscope Kit EW-87701-24 Mfr # 6210-021
COMPARE
6-digit numeric, 5-digit alphanumeric backlit LCD 8 to 20 µsec
$1,155.00USD / Each
Showing 1- 7 of 7