Noshock 100 Series Current Output Pressure Transducers
Current Output Pressure Transmitters
  • Accuracy up to ±0.25 % Full Scale (Best Fit Straight Line)
  • Welded stainless steel pressure chamber
  • Advanced diffused semiconductor and sputtered thin film sensor for maximum stability
  • Compact size
  • High alternating load resistance
  • High overpressure protection
  • CE compliant to suppress RFI, EMI and ESD
  • Compatible with NOSHOK Smart System Indicators
NOSHOK 100 Series Current Output Pressure Transmitters are designed to provide a previously unequalled level of performance, utilizing diffused semiconductor and sputtered thin film strain gauge technology. 100 Series transmitters are highly repeatable, shock resistant and are extremely stable over long periods of time. CE compliance which includes substantial levels of RFI, EMI and ESD protection combined with reverse polarity and over-voltage protection insure they perform well in the most demanding applications. Advanced manufacturing techniques combined with technologically advanced standard features allow NOSHOK to offer a level of performance previously found only on transducers costing hundreds of dollars more. Final calibration tests performed on all NOSHOK transmitters prior to sh
ITEM(S) ON SALE
$509.75 - $687.73USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
NOSHOK 100-30/15-1-1-2-7 Transmitters, -30 INHG TO 15 PSIG, 4-20MA, 1/4" NPT MALE EW-68028-52 Mfr # 100-30/15-1-1-2-7
COMPARE
±0.5 % full scale 15
$687.73USD / Each
$731.56USD / Each
NOSHOK 100-15-1-1-2-7 0 PSIG To 15 PSIG, 4-20 mA, 1/4" NPT Male EW-68028-61 Mfr # 100-15-1-1-2-7
COMPARE
±0.5 % full scale 15
$509.75USD / Each
$539.26USD / Each
NOSHOK 100-30-1-1-2-7 0 PSIG To 30 PSIG, 4-20 mA, 1/4" NPT Male EW-68028-62 Mfr # 100-30-1-1-2-7
COMPARE
±0.5 % full scale 30
$509.75USD / Each
$539.26USD / Each
NOSHOK 100-60-1-1-2-7 0 PSIG To 60 PSIG, 4-20 mA, 1/4" NPT Male EW-68028-63 Mfr # 100-60-1-1-2-7
COMPARE
±0.5 % full scale 60
$509.75USD / Each
$539.79USD / Each
NOSHOK 100-100-1-1-2-7 0 PSIG To100 PSIG, 4-20 mA, 1/4" NPT Male EW-68028-64 Mfr # 100-100-1-1-2-7
COMPARE
±0.5 % full scale 100
$509.75USD / Each
$539.26USD / Each
NOSHOK 100-600-1-1-2-7 0 PSIG To 600 PSIG, 4-20 mA, 1/4" NPT Male EW-68028-69 Mfr # 100-600-1-1-2-7
COMPARE
±0.5 % full scale 600
$509.75USD / Each
$539.26USD / Each
NOSHOK 100-5000-1-1-2-7 0 PSIG To 5000 PSIG, 4-20 mA, 1/4" NPT Male EW-68028-75 Mfr # 100-5000-1-1-2-7
COMPARE
±0.5 % full scale 5000
$509.75USD / Each
$588.37USD / Each
Showing 1- 7 of 7