MYRON L® Conductivity Calibration Solutions
Conductivity values are accurate to within ±1% and are referenced to 25°C
  • Standards have NaCl and 442™ conversions included for ease of use
$55.30 - $64.00USD / Quart of 1
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Conductivity Value (µS) Description
Availability Pricing
Myron L 15 QCP (NIST) conductivity calibration standard, 23.8 µS
Always in Stock
EW-01489-90 Mfr # 442-15Q-CP
COMPARE
23.8 conductivity calibration standard, 23.8 µS
$61.90USD / Quart of 1
Myron L 30 QCP (NIST) conductivity calibration standard, 46.7 µS
Always in Stock
EW-01489-91 Mfr # 442-30Q-CP
COMPARE
46.7 conductivity calibration standard, 46.7 µS
$64.00USD / Quart of 1
Myron L 70 QCP (NIST) conductivity calibration standard, 70 µS
Always in Stock
EW-01489-92 Mfr # KCL-70Q-CP
COMPARE
70 conductivity calibration standard, 70 µS
$61.30USD / Quart of 1
Myron L 300 QCP (NIST) conductivity calibration standard, 445 µS
Always in Stock
EW-01489-93 Mfr # 442-300Q-CP
COMPARE
445 conductivity calibration standard, 445 µS
$55.30USD / Quart of 1
Myron L 700 QCP (NIST) conductivity calibration standard, 700 µS
Always in Stock
EW-01489-94 Mfr # KCL-700Q-CP
COMPARE
700 conductivity calibration standard, 700 µS
$56.40USD / Quart of 1
Myron L 1500 QCP (NIST) conductivity calibration standard, 2060 µS
Always in Stock
EW-01489-95 Mfr # 442-1500Q-CP
COMPARE
2060 conductivity calibration standard, 2060 µS
$56.40USD / Quart of 1
Myron L 3K QCP (NIST) conductivity calibration standard, 3900 µS
Always in Stock
EW-01489-96 Mfr # 442-3000Q-CP
COMPARE
3900 conductivity calibration standard, 3900 µS
$56.40USD / Quart of 1
Myron L 7K QCP (NIST) conductivity calibration standard, 7000 µS EW-01489-97 Mfr # KCL-7000Q-CP
COMPARE
7000 conductivity calibration standard, 7000 µS
$56.40USD / Quart of 1
Myron L 15K QCP (NIST) conductivity calibration standard, 16,630 µS
Always in Stock
EW-01489-98 Mfr # 442-15000Q-CP
COMPARE
16630 conductivity calibration standard, 16,630 µS
$61.20USD / Quart of 1
Myron L 30K QCP (NIST) conductivity calibration standard, 30,100 µS
Always in Stock
EW-01489-99 Mfr # 442-30000Q-CP
COMPARE
30100 conductivity calibration standard, 30,100 µS
$57.50USD / Quart of 1
Showing 1- 10 of 10