Multiwash III Plate Washers
  • Quick-connect fittings allow unit to be fully operational in 15 minutes
  • Rinse-on-demand feature for complete system cleaning at the touch of a button
  • Can handle full range of assay types from the most gentle wash to the most aggressive ELISA
These plate washers feature easy-to-program wash routines, bottom washing, sweeping crosswise aspiration, dispense only and aspirate only functions, and overflow washing for continual dispensing while aspirating off the top for a thorough wash. Use the plate washer to perform cell washing; agglutination tests; cytotoxicity tests; and ELISA, EIA, FIA, and LIA tests.
Minimum residual volume is ≤1 μL per well. Washer has seven programmable wash operations. Independent adjustments of horizontal and vertical needle positions allow any shape well to be accommodated; extra aspiration adjustment for both horizontal and vertical needle positions can be stored in individual programs. Wetted materials include glass, nylon, PTFE, stainless steel, polypropylene, Tygon®, Delrin®, polyethylene, and Santoprene®.
The washer performs a 300-µL three-cycle wash on a full 96-well plate in less than two minutes. You control the dispense pressure. By programming and storing routines, you can switch between seven different modes at the touch of a button. The washer also features a floating manifold that adjusts to match each plate and allows the washer to perform special test procedures. All units are CE-approved, ETL-listed, and certified with CAN/CSA Std C22.2, No. 1010.1.
$685.50 - $9,653.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Accuracy Number of Channels (Manifold) Description
Availability Pricing
MultiWash III Plate Washer with 8-Port Manifold EW-13054-02 Mfr # 820-01(MULTIWASH3)
COMPARE
±3% at 300 μL 8 Plate Washer with 8-Port Manifold
$9,653.00USD / Each
MultiWash III Plate Washer with 8-Port Coaxial Manifold EW-13054-14 Mfr # 820-04(MULTIWASH3)
COMPARE
±3% at 300 μL 8 Plate Washer with 8-Port Coaxial Manifold
$8,542.00USD / Each
COMPARE
±3% at 300 μL 8 8-Port Standard Manifold for III Plate Washers
$685.50USD / Each
COMPARE
±3% at 300 μL 8 8-Port Coaxial Manifold for III Plate Washers
$707.50USD / Each
COMPARE
±3% at 300 μL 12 12-Port Coaxial Manifold for III Plate Washers
$991.00USD / Each