This product is now discontinued, View Related Products
Motic Compound Microscopes with Digital Camera
Color-corrected infinity optical system for high-image quality and flatness of field
  • Stream live images to your PC
Ideal for use with life sciences, biological applications, hematology, cytology in universities and laboratories. View and download images through the USB port. Powerful software tools make image analysis easy: view live images on your PC; accurately measure and quantify your specimen and export results in an Excel® format for analysis; count and evaluate cell quantity with the Auto Segmentation and Calculation tool; combine two images to bring out differences; add text, symbols, and even sound recordings to your image.
Microscopes feature eyepieces with a high eye point for eyeglass wearers and diopter adjustment for both eyes. The observation tubes have a 30° viewing angle and interpupillary adjustment of 55 to 75 mm.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Illuminator(s) Maximum Magnification Stage Dimensions
Availability Pricing
Motic Instruments 1100401200482 Compound Microscope with Digital Camera EW-03911-40 Mfr # 1100401200615
COMPARE
Halogen with rheostat, 30 W 1000x 140 x 135 mm
Call For Price
Motic Instruments 1100401200492 Compound Microscope with Digital Camera, Koehler illumination EW-03911-45 Mfr # 1100401200492
COMPARE
Koehler halogen with rheostat, 30 W 1000x 160 x 140 mm