Monarch Instrument Optical Sensors

Choose from 5 sensor options for a wide range of applications
  • Optical sensor for speed measurments
  • Order tachometer panel meter separately for speed measurment readings
Proximity Sensor is a probe-style inductive sensor for use up to 0.2" (0.5 cm) from a 0.5" (1.3 cm) diameter metallic target.
Magnetic Sensor with amplifier is for use with 60 tooth, 20 pitch gear.
Remote Optical Sensor is a stainless steel sensor with visible red LED light source and green LED on-target indicator. For distances up to 3 ft (1 m), up to 45° off center from reflective target.
Infrared Sensor is for use with or without reflective tape at 0.5" (1.3 cm) sensing gap from target. Senses white to black contrasting marks, or beam can be interrupted by solid objects.
Remote Optical Laser Sensor is ideal for a wide range of applications where distance to target is large.
$120.80 - $220.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Measuring Distance (in) Max RPM Range (rpm) Memory
Availability Pricing
Monarch Instrument MI28905 Proximity sensor for 1/8 DIN panel-mount tachometers EW-08212-52 Mfr # 6180-013
COMPARE
0.2 60000
$120.80USD / Each
Monarch 6180-056 Optical Sensor, for up to 3 feet. Range from 1 to 250,000 rpm EW-08212-53 Mfr # 6180-056
COMPARE
$199.00USD / Each
Monarch Instrument 6180-012 M-190WMagnetic Sensor W/ 8ft Cable () EW-08212-56 Mfr # 6180-012
COMPARE
100000
$126.00USD / Each
Monarch 6180-021 IRS-W Infrared Sensor with 8ft Cable EW-98806-77 Mfr # 6180-021
COMPARE
$220.00USD / Each
Monarch ROS-P-25 : REMOTE OPTICAL SENS EW-98806-82 Mfr # 6180-057-025
COMPARE
$175.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat