Monarch Instrument Nova-Strobe LED Portable Stroboscopes
Adjustable brightness control in a lightweight Industrial design
  • Twelve button keypad makes entering flash rates quick and easy (DBL, PBL models)
  • Energy efficient with long battery life – up to 10 hours
These rugged industrial LED stroboscopes provide an extremely bright, uniform light output for performing stop motion diagnostic inspection and RPM measurements. The twelve LED light source design provides an extremely efficient light source resulting in continuous cool operations leading to a long battery life. With a wide operating range of 30 to 500,000 flashes per minute these units cover most applications.
The BBL Basic model is designed for simple stop motion inspection and RPM measurement applications. The DBL Deluxe model includes all standard features of the BBL Basic with the addition of internal phase shifting, memory for up to five preset flash rates, N.I.S.T. calibration certificate, and tachometer mode for speed measurements up to 500,000 rpm using optional remote sensors or TTL pulse input/output. The PBL Phaser advanced model has all the features of the DBL with the addition of external phase delay, time delay, virtual rpm mode, and a power supply/recharger that allows for 24/7 continuous operation.
$781.00 - $1,330.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Memory Power (VAC) Description
Availability Pricing
Monarch 6230-010 Nova-Strobe BBL LED Stroboscope EW-53502-00 Mfr # 6230-010
COMPARE
N/A Nova-Strobe BBL LED Stroboscope
$781.00USD / Each
Monarch 6230-011 Nova-Strobe BBL LED Stroboscope Kit EW-53502-01 Mfr # 6230-011
COMPARE
N/A Nova-Strobe BBL LED Stroboscope Kit
$906.00USD / Each
Monarch 6231-010 Nova-Strobe DBL LED Stroboscope EW-53502-02 Mfr # 6231-010
COMPARE
Programmable Nova-Strobe DBL LED Stroboscope
$918.50USD / Each
Monarch 6231-011 Nova-Strobe DBL LED Stroboscope Kit EW-53502-03 Mfr # 6231-011
COMPARE
Programmable Nova-Strobe DBL LED Stroboscope Kit
$1,064.00USD / Each
Monarch 6232-010 Nova-Strobe PBL LED Stroboscope EW-53502-04 Mfr # 6232-010
COMPARE
Programmable Nova-Strobe PBL LED Stroboscope
$1,188.00USD / Each
Monarch 6232-011 Nova-Strobe PBL LED Stroboscope Kit EW-53502-05 Mfr # 6232-011
COMPARE
Programmable Nova-Strobe PBL LED Stroboscope Kit
$1,330.00USD / Each