Monarch Instrument 2 Channel Paperless Recording Systems
The DataChart 1250 is a powerful, compact digital recorder with intuitive operation, flexible input configuration and large memory capability. Combining small package size with extreme flexibility make the DataChart 1250 a powerful, yet economical, digital data recording system.
FEATURES
  • 2 universally configurable isolated inputs for measuring DC voltage, DC current, thermocouples, RTD's as well as frequency and pulse inputs
  • 4 internal alarm set points
  • 2 alarm relay outputs and 1 digital control input
  • Maximum sample storage rate of 100 samples per second per channel
  • Compact Flash cards up to 2 Gigabyte size
  • $943.00USD / Each
    1 variations of this product are available.
    See All Product Options Below
    1 PRODUCT OPTIONS
    Compareitem specifications
    Chart Diameter (in) Chart Speed Max Input (VDC)
    Availability Pricing
    Monarch DC1250-U10 Recorder; Universal AC Adapter/Rechargeable EW-49005-91 Mfr # DC1250-U10
    COMPARE
    $943.00USD / Each