MLA Precision Model Pipettes
Ideal for a broad range of laboratory procedures requiring precision and accuracy
  • Choose from fixed volumes between 5 µL up to 1000 µL
  • Simple single-stroke procedure
  • Lower spring force minimizes muscle aches due to repetitive motion
The MLA Precision pipettes are calibrated “to deliver” (TD) pipettes, use air-displacement, and require manual de-tipping. They come factory calibrated and tested at 21.5°C ±2°C using distilled water. Pipettors consistently deliver the calibrated volume whenever the plunger is fully depressed. Calibration volume can be changed using the calibration key (included).
Pipettors are color-coded for easy identification and come with the stated volume engraved onto the piston assembly. Engineered with aluminum alloy and stainless steel, these pipettors withstand years of use and provide exceptional performance for a wide range of fluids, including dense or viscous samples.
$234.00 - $248.50USD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Color Code Max Volume (µL)
Availability Pricing
VistaLab 1021 Fixed-Volume Pipette, Silver, 10 uL; Each EW-06197-55 Mfr # 1021
COMPARE
±5.00% Orange 10
$234.00USD / Each
VistaLab 1022 Fixed-Volume Pipette, Black, 20 uL; Each EW-06197-57 Mfr # 1022
COMPARE
±3.00% Black 20
$234.00USD / Each
VistaLab 1023 Fixed-Volume Pipette, White, 25 uL; Each EW-06197-59 Mfr # 1023
COMPARE
±3.00% White 25
$234.00USD / Each
VistaLab 1024 Fixed-Volume Pipette, Green, 50 uL; Each EW-06197-61 Mfr # 1024
COMPARE
±1.00% Green 50
$234.00USD / Each
VistaLab 1025 Fixed-Volume Pipette, Blue, 100 uL; Each EW-06197-63 Mfr # 1025
COMPARE
±1.00% Blue 100
$234.00USD / Each
VistaLab 1026 Fixed-Volume Pipette, Red, 200 uL; Each EW-06197-65 Mfr # 1026
COMPARE
±1.00% Red 200
$234.00USD / Each
VistaLab 1027 Fixed-Volume Pipette, Purple, 250 uL; Each EW-06197-69 Mfr # 1027
COMPARE
±1.00% Purple 250
$234.00USD / Each
VistaLab 1028 Fixed-Volume Pipette, Brown, 300 uL; Each EW-06197-91 Mfr # 1028
COMPARE
±1.00% Brown 300
$234.00USD / Each
VistaLab 1029 Fixed-Volume Pipette, Yellow, 500 uL; Each EW-06197-93 Mfr # 1029
COMPARE
±1.00% Yellow 500
$234.00USD / Each
VistaLab 1030 Fixed-Volume Pipette, Gray, 1000 uL; Each EW-06197-95 Mfr # 1030
COMPARE
±1.00% Gray 1000
$234.00USD / Each
VistaLab 1101 Fixed-Volume Pipette, Silver, 5 uL; Each EW-06197-97 Mfr # 1101
COMPARE
±5.00% Silver 5
$234.00USD / Each
VistaLab 1102 Fixed-Volume Pipette, Silver, 15 uL; Each EW-06197-99 Mfr # 1102
COMPARE
±5.00% Silver 15
$234.00USD / Each
VistaLab 1103 Fixed-Volume Pipette, Silver, 30 uL; Each EW-06198-01 Mfr # 1103
COMPARE
±3.00% Silver 30
$248.50USD / Each
VistaLab 1104 Fixed-Volume Pipette, Silver, 40 uL; Each EW-06198-03 Mfr # 1104
COMPARE
±3.00% Silver 40
$234.00USD / Each
VistaLab 1106 Fixed-Volume Pipette, Silver, 60 uL; Each EW-06198-07 Mfr # 1106
COMPARE
±2.00% Silver 60
$234.00USD / Each
VistaLab 1108 Fixed-Volume Pipette, Silver, 75 uL; Each EW-06198-11 Mfr # 1108
COMPARE
±2.00% Silver 75
$234.00USD / Each
VistaLab 1111 Fixed-Volume Pipette, Silver, 120 uL; Each EW-06198-17 Mfr # 1111
COMPARE
±1.00% Silver 120
$234.00USD / Each
VistaLab 1113 Fixed-Volume Pipette, Silver, 400 uL; Each EW-06198-21 Mfr # 1113
COMPARE
±1.00% Silver 400
$234.00USD / Each
VistaLab 1118 Fixed-Volume Pipette, Silver, 900 uL; Each EW-06198-27 Mfr # 1118
COMPARE
±1.00% Silver 900
$234.00USD / Each
VistaLab 1119 Fixed-Volume Pipette, Silver, 450 uL; Each EW-06198-29 Mfr # 1119
COMPARE
±1.00% Silver 450
$234.00USD / Each
Showing 1- 20 of 20