Mini-Dial Durometers
Meet your specific application
  • Compliance: ASTM D-2240
Mini-Dial Durometers are compact and easy to carry around the plant or take in the field for on-site testing. The small dial face measures only 1-5/8". Scale is divided into 1 point increments from 0 to 100. The durometer holds maximum reading until reset. Ideal for testing hard-to-reach places or for out-of-sight testing. Measures 1-5/8"W x 5-5/8"H.
$575.50 - $613.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Calibration Included Type
Availability Pricing
Rex Gauge H-1000 TYPE A Mini-Dial Hardness Tester/Durometer; Type A EW-59792-04 Mfr # H-1000 TYPE A
COMPARE
No A
$587.50USD / Each
Rex Gauge H-1000 TYPE D Mini-Dial Hardness Tester/Durometer; Type D EW-59792-06 Mfr # H-1000 TYPE D
COMPARE
No D
$607.50USD / Each
Rex Gauge H-1000 TYPE A Mini-Dial Durometer with NIST Calibration Certificate, Type A EW-59792-32 Mfr # H-1000 TYPE A W/NIST CERT
COMPARE
Yes A
$613.00USD / Each
Rex Gauge H-1000 TYPE D Mini-Dial Durometer with NIST Calibration Certificate, Type D EW-59792-34 Mfr # H-1000 TYPE D W/NIST CERT
COMPARE
Yes D
$575.50USD / Each