Micro A07 Citric Acid Cleaner
Environmentally friendly – Phosphate Free
  • Free Rinsing
Micro A07 is a free-rinsing cleaner designed for use in a wide range of applications including ultrasonic baths, immersion tanks, mild-agitation systems, CIP systems and more. It is ideal for cleaning metals, stainless steel, ceramics, glass, plastics, and filter membranes. Micro A07 solution removes a variety of soils as well including scale, oxides, milkstone, mineral deposits, and inorganic materials. It is free of CFCs, ODCs, phosphates, silicates, borates, halogenated compounds, and phenols.
$35.50 - $3,360.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Format Volume (Liters) Description
Availability Pricing
M-0890 Micro A07 Citric Acid Liquid Cleaner, 225 KG EW-18100-07 Mfr # M-0890
COMPARE
Liquid M-0890 MICRO A07 CITRIC ACID CLEANER 225 KG NET WT PLASTIC DRUM
$3,360.00USD / Each
International Products Corp M-0801-12 Micro A07 Citric Acid Liquid Cleaner, 1 Liter EW-18104-00 Mfr # M-0801-12
COMPARE
Liquid 1 Micro A07 Citric Acid Liquid Cleaner, 1 Liter
$35.50USD / Each
International Products Corp M-0801-12 Micro A07 Citric Acid Cleaner, 12/cs EW-18104-01 Mfr # M-0801-12
COMPARE
Liquid 1 Micro A07 Citric Acid Cleaner case of 12 1L bottles
$385.00USD / Case of 1