Mettler Toledo XPR Micro Balances/Mass Comparators
Perform unrivaled weighing accuracy and repeatability
  • One-click calibration certificate
  • USB and Ethernet interfaces easily transfers data to peripheral devices or network
  • Large color touchscreen with built-in user guidance simplifies operation
  • MC Link calibration software automatically stores all weight calibrations
  • StatusLight™ uses colored lights to indicate when balance is ready for weighing
The XPR micro-analytical mass comparator balances offer you one of the highest weighing accuracies for determination of mass up to 200 g. The hanging weighing pan eliminates corner load effects, eccentricity errors, and makes it easy to place weights and samples in the correct position.
The MC Link software stores all your weight calibration data and other relevant metrological data in a secure database. Calibration certificates can be easily issued with one click. Integrated USB and Ethernet interfaces allow you quickly transfer data to peripheral devices or network. Data can also be transferred to MC Link or third-party software.
Special features include built-in ProFACT adjustment, a large touchscreen terminal with user guidance, automatic static detection, and intelligent quality assurance functions, all designed to simplify your processes and eliminate errors.
$23,870.00 - $47,140.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Mettler Toledo XPR36C Micro Balance / Mass Comparator, 32 g x 0.001 mg, Motorized Draft Shield EW-11334-43 Mfr # XPR36C
COMPARE
Internal 32 0.001
$38,880.00USD / Each
Mettler Toledo XPR226CDR Micro Balance / Mass Comparator, 121/220 g x 0.01/0.1 mg, Motorized Draft Shield EW-11334-45 Mfr # XPR226CDR
COMPARE
Internal 220 0.005
$23,870.00USD / Each
Mettler Toledo XPR206CDR Micro Balance/Mass Comparator, 220 g/121 g x 0.01/0.005 mg , Motorized Draft Shield EW-11335-21 Mfr # XPR206CDR
COMPARE
Internal 220 0.01
$24,640.00USD / Each
Mettler Toledo XPR26C Micro Balance/Mass Comparator, 22 g x 0.001 mg, Motorized Draft Shield EW-11335-23 Mfr # XPR26C
COMPARE
Internal 22 0.001
$40,130.00USD / Each
Mettler Toledo XPR56C Micro Balance/Mass Comparator, 52 g x 0.001 mg, Motorized Draft Shield EW-11335-25 Mfr # XPR56C
COMPARE
Internal 52 0.001
$47,140.00USD / Each