Mettler Toledo NewClassic MS-TS Toploading Balances
Fast settling time and consistent results for routine to complicated weighing procedures
  • Extra-large color TFT touchscreen display with intuitive user interface
  • Touchscreen can be operated with cotton, silicon and rubber gloves
  • LevelControl warning the user when the balance is not level and provides onscreen help to correct
  • Advanced MonoBloc weighing technology provides fast and accurate results
  • SmartTrac software displays how much of the entire weighing range has been used
  • MinWeigh function displays the weight value in red until the net sample is above the pre-programmed minimum value
  • Passcodes restrict modification of the balance settings to authorized personnel only
  • ISOLog function maintains a record of all weighing relevant changes to the balance’s settings
  • Fully automatic time- and temperature-controlled internal calibration adjustment
  • Durable high-grade chemical-resistant die-cast aluminum housing
  • Overload Protection guards the weighing cell against excess weight overload
  • Direct connection to a PC or printer via RS232 or USB slot allows convenient transfer of data
The high capacity MS balances 12,200 g and above come with a high-contrast display (HCD) with tactial buttons. These balance do not include touchscreen, LevelControl, MinWeigh, passcodes or ISOLog features.
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Mettler Toledo MS303TS Newclassic MS-TS Toploading Balance with Shield, 320g x 1 Mg EW-11335-47 Mfr # MS303TS
COMPARE
320 100 to 240 0.001
$4,755.00USD / Each
Mettler Toledo MS403TS Newclassic MS-TS Toploading Balance with Shield, 420g x 1 Mg EW-11335-48 Mfr # MS403TS
COMPARE
420 100 to 240 0.001
$5,255.00USD / Each
Mettler Toledo MS603TS Newclassic MS-TS Toploading Balance with Shield, 620g x 1 Mg EW-11335-49 Mfr # MS603TS
COMPARE
620 100 to 240 0.001
$6,200.00USD / Each
Mettler Toledo MS1003TS Newclassic MS-TS Toploading Balance with Shield, 1020g x 1 Mg EW-11335-50 Mfr # MS1003TS
COMPARE
1020 100 to 240 0.001
$7,160.00USD / Each
Mettler Toledo MS1602TS Newclassic MS-TS Toploading Balance, 1620g x 0.01g EW-11335-51 Mfr # MS1602TS
COMPARE
1620 100 to 240 0.01
$4,345.00USD / Each
Mettler Toledo MS3002TS Newclassic MS-TS Toploading Balance, 3200g x 0.01g EW-11335-52 Mfr # MS3002TS
COMPARE
3200 100 to 240 0.01
$5,095.00USD / Each
Mettler Toledo MS4002TS Newclassic MS-TS Toploading Balance, 4200g x 0.01g EW-11335-53 Mfr # MS4002TS
COMPARE
4200 100 to 240 0.01
$5,395.00USD / Each
Mettler Toledo MS4002TSDR Newclassic DeltaRange Toploading Balance, 4200/820G x 0.1/0.01g EW-11335-54 Mfr # MS4002TSDR
COMPARE
4200 100 to 240 0.1
$4,710.00USD / Each
Mettler Toledo MS6002TS Newclassic MS-TS Toploading Balance, 6200g x 0.01g EW-11335-55 Mfr # MS6002TS
COMPARE
6200 100 to 240 0.1
$6,330.00USD / Each
Mettler Toledo MS8001TS Newclassic MS-TS DeltaRange Toploading Balance, 8200g x 0.1g EW-11335-57 Mfr # MS8001TS
COMPARE
8200 100 to 240 0.1
$4,700.00USD / Each
Mettler Toledo MS12002TS Newclassic MS-TS Toploading Balance, 12200g x 0.01g EW-11335-58 Mfr # MS12002TS
COMPARE
12200 100 to 240 0.01
$7,590.00USD / Each
Showing 1- 11 of 11