Mettler Toledo NewClassic ML-T Precision Toploading Balances
Designed for mobility and easy operation
  • Extra-large color TFT touch-screen display with intuitive user interface
  • Touch-screen can be operated with cotton, silicone, and rubber gloves
  • AC or battery operable for up to 8 hours—ideal for work in field or whenever a power supply is hard to find
  • LevelControl warns you when the balance is not level and offers onscreen help
  • Advanced MonoBloc weighing technology provides fast and accurate results
  • MinWeigh function displays the weight value in red until the net sample is above the preprogrammed minimum value
  • Pass codes restrict modification of the balance settings to authorized personnel only
  • Overload protection guards the weighing cell against excess weight overload
  • Direct connection to a PC or printer via RS-232 or USB slot allows convenient transfer of data
  • Easy-clean feature saves time and effort with all washable parts being dishwasher-safe
These compact precision balances allow you to take them where they are needed most and offer extra space for all your other equipment and samples. Ideal for stocktaking or fieldwork. The 4.5" color TFT touchscreen features clear icons making it easy to operate quickly even while wearing gloves.
Built-in overload protection ensures a long balance lifetime. The MonoBloc weighing cell, with FACT automatic internal adjustment, delivers consistently reliable results.
ITEM(S) ON SALE
$3,300.00 - $5,340.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Mettler Toledo ML203T Newclassic Precision Toploading Balance; 220 g x 1 mg EW-11337-04 Mfr # ML203T
COMPARE
220 100 to 240 0.001
$3,920.00USD / Each
Mettler Toledo ML303T Newclassic Precision Toploading Balance; 320 g x 1 mg EW-11337-05 Mfr # ML303T
COMPARE
320 100 to 240 0.001
$4,115.00USD / Each
Mettler Toledo ML503T Newclassic Precision Toploading Balance; 520 g x 1 mg EW-11337-06 Mfr # ML503T
COMPARE
520 100 to 240 0.001
$4,855.00USD / Each
Mettler Toledo ML802T Newclassic Precision Toploading Balance; 820 g x 0.01 g EW-11337-07 Mfr # ML802T
COMPARE
820 100 to 240 0.01
$3,300.00USD / Each
Mettler Toledo ML1602T Newclassic Precision Toploading Balance; 1620 g x 0.01 g EW-11337-08 Mfr # ML1602T
COMPARE
1620 100 to 240 0.01
$3,700.00USD / Each
Mettler Toledo ML3002T Newclassic Precision Toploading Balance; 3200 g x 0.01 g EW-11337-45 Mfr # ML3002T
COMPARE
3200 100 to 240 0.01
$4,335.00USD / Each
Mettler Toledo ML4002T Newclassic Precision Toploading Balance; 4200 g x 0.01 g EW-11337-46 Mfr # ML4002T
COMPARE
4200 100 to 240 0.01
$4,575.00USD / Each
Mettler Toledo ML6002T Newclassic Precision Toploading Balance; 6200 g x 0.01 g EW-11337-47 Mfr # ML6002T
COMPARE
6200 100 to 240 0.01
$5,340.00USD / Each
Mettler Toledo ML3001T Newclassic Precision Toploading Balance; 3200 g x 0.1 g EW-11337-48 Mfr # ML3001T
COMPARE
3200 100 to 240 0.1
$3,595.00USD / Each
Mettler Toledo ML6001T Newclassic Precision Toploading Balance; 6200 g x 0.1 g EW-11337-49 Mfr # ML6001T
COMPARE
6200 100 to 240 0.1
$3,635.45USD / Each
$3,995.00USD / Each
Showing 1- 10 of 10