Mettler Toledo® NewClassic ME-T Toploading Balances
Easy worry-free weighing that delivers consistently reliable results
  • Large 4.5" color TFT touchscreen and intuitive interface simplify operation
  • Electromagnetic Force Compensation (EMFC) weighing cell with Fully Automatic Calibration Technology (FACT) automatically adjusts the balance to ensure accuracy
  • Internal calibration provides simple and reliable use
  • Ten embedded applications include essential weighing functions, statistics, and formulation
  • Sample ID and passcode protection provide setting protections and produce GLP documentation
  • RS-232 and USB connectivity simplify data upload and transfer
The Mettler Toledo® ME-T toploading balances are ideal for routine laboratory weighing. They feature a 4.5” color TFT touchscreen with intuitive guidance enabling you to operate quickly and efficiently, even while wearing gloves. An electronmagnetic force compensation (EMFC) weighing cell with fully automated calibration technology (FACT) automatically adjusts the balance to provide consistent reliable results. Multiple interfaces support barcode reader, printer and PC connections for easy traceability and data handling. USB and RS-232 connectivity along with an optional Bluetooth adapter simplify data upload and transfer. Bluetooth adapter sold separately as part number 11140-79. Constructed with a durable metal base housing and high-quality finish, these balances are designed for tough environments while being easy to maintain and clean.
ITEM(S) ON SALE
$2,297.75 - $3,418.87USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Mettler Toledo ME103T NewClassic ME-T Precision Toploading Balance; 120 g x 1 mg EW-11348-03 Mfr # ME103T
COMPARE
Internal 120 0.001
$2,366.00USD / Each
$2,600.00USD / Each
Mettler Toledo ME203T NewClassic ME-T Precision Toploading Balance; 220 g x 1 mg EW-11348-04 Mfr # ME203T
COMPARE
Internal 220 0.001
$2,820.00USD / Each
Mettler Toledo ME303T NewClassic ME-T Precision Toploading Balance; 320 g x 1 mg EW-11348-05 Mfr # ME303T
COMPARE
Internal 320 0.001
$2,980.00USD / Each
Mettler Toledo ME403T NewClassic ME-T Precision Toploading Balance; 420 g x 1 mg EW-11348-06 Mfr # ME403T
COMPARE
Internal 420 0.001
$2,948.40USD / Each
$3,240.00USD / Each
Mettler Toledo ME1002T NewClassic ME-T Precision Toploading Balance; 1200 g x 10 mg EW-11348-07 Mfr # ME1002T
COMPARE
Internal 1200 0.01
$2,366.00USD / Each
$2,600.00USD / Each
Mettler Toledo ME2002T NewClassic ME-T Precision Toploading Balance; 2200 g x 10 mg EW-11348-08 Mfr # ME2002T
COMPARE
Internal 2200 0.01
$2,865.00USD / Each
Mettler Toledo ME3002T NewClassic ME-T Precision Toploading Balance; 3200 g x 10 mg EW-11348-09 Mfr # ME3002T
COMPARE
Internal 3200 0.01
$2,821.00USD / Each
$3,100.00USD / Each
Mettler Toledo ME4002T NewClassic ME-T Precision Toploading Balance; 4200 g x 10 mg EW-11348-10 Mfr # ME4002T
COMPARE
Internal 4200 0.01
$2,993.90USD / Each
$3,290.00USD / Each
Mettler Toledo ME4001T NewClassic ME-T Precision Toploading Balance; 4200 g x 100 mg EW-11348-11 Mfr # ME4001T
COMPARE
Internal 4200 0.1
$2,297.75USD / Each
$2,525.00USD / Each
Mettler Toledo ME503T NewClassic ME-T Precision Toploading Balance; 520 g x 1 mg EW-11348-12 Mfr # ME503T
COMPARE
Internal 520 0.001
$3,410.00USD / Each
Mettler Toledo ME5002T NewClassic ME-T Precision Toploading Balance; 5200 g x 10 mg EW-11348-13 Mfr # ME5002T
COMPARE
Internal 5200 0.01
$3,171.35USD / Each
$3,485.00USD / Each
Mettler Toledo ME603T/00 NewClassic ME-T Precision Toploading Balance, 620 g x 1 mg EW-55000-57 Mfr # 30704481
COMPARE
Internal 620 0.001
$3,418.87USD / Each
$3,757.00USD / Each
Mettler Toledo ME6002T/00 NewClassic ME-T Precision Toploading Balance, 6200 g x 10 mg EW-55000-59 Mfr # 30704479
COMPARE
Internal 6200 0.01
$3,418.87USD / Each
$3,757.00USD / Each
Showing 1- 13 of 13