Mettler Toledo SmartCal Moisture Analyzer Test Substances
Quick and Easy Performance Test for Moisture Analyzers
  • Indicates incorrect performance immediately
  • Test temperatures include 70, 100, 130, 160°C (158, 212, 266, 320°F)
SmartCal verifies the performance of your moisture analyzer in a 10-minute test, which is carried out in the same easy way as a regular measurement. Regular testing with SmartCal lets you provides documented evidence of the unit's performance over time. Incorrect performance is immediately indicated. The test instantly indicates adverse influences from the heating or weighing units. If the test is passed, you can trust your results.
ITEM(S) ON SALE
$476.43 - $1,285.00USD / Pkg of 24
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Accessory Type Description
Availability Pricing
Mettler Toledo cSmartCal24 Moisture Analyzer Reference Substance with Certificate; 24 Tests EW-11336-29 Mfr # 30005791
COMPARE
For use with Mettler Toledo Halogen Moisture Analyzer SmartCal Moisture Analyzer Reference Substance with Certificate; 24 Tests
$1,285.00USD / Pkg of 24
Mettler Toledo cSmartCal12 Moisture Analyzer Reference Substance with Certificate; 12 Tests EW-11336-30 Mfr # 30005793
COMPARE
For use with Mettler Toledo Halogen Moisture Analyzer SmartCal Moisture Analyzer Reference Substance with Certificate; 12 Tests
$697.81USD / Pkg of 12
$725.00USD / Pkg of 12
Mettler Toledo SmartCal24 Moisture Analyzer Reference Substance; 24 Tests EW-11336-31 Mfr # 30005790
COMPARE
For use with Mettler Toledo Halogen Moisture Analyzer SmartCal Moisture Analyzer Reference Substance; 24 Tests
$856.62USD / Pkg of 24
$890.00USD / Pkg of 24
Mettler Toledo SmartCal12 Moisture Analyzer Reference Substance; 12 Tests EW-11336-32 Mfr # 30005792
COMPARE
For use with Mettler Toledo Halogen Moisture Analyzer SmartCal Moisture Analyzer Reference Substance; 12 Tests
$476.43USD / Pkg of 12
$495.00USD / Pkg of 12
Mettler Toledo cSmartCal StarterPac Moisture Analyzer Reference Substance with Certificate; 12 Tests EW-11336-33 Mfr # 30005918
COMPARE
For use with Mettler Toledo Halogen Moisture Analyzer SmartCal StarterPac Moisture Analyzer Reference Substance with Certificate; 12 Tests
$810.00USD / Pkg of 12
Mettler Toledo SmartCal StarterPac  Moisture Analyzer Reference Substance, 12 Tests EW-11336-34 Mfr # 30005917
COMPARE
For use with Mettler Toledo Halogen Moisture Analyzer SmartCal StarterPac Moisture Analyzer Reference Substance, 12 Tests
$595.00USD / Pkg of 12