This product is now discontinued, View Related Products

Fisher Chemical™ Methyl Ethyl Ketone, ACS Reagent Grade

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

10 variations of this product are available.

See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Fisher Chemical Methyl Ethyl Ketone, ACS Reagent Grade, 500 mL EW-88016-50 Mfr # M209500
COMPARE
78-93-3 500 Methyl Ethyl Ketone, ACS Reagent Grade, 500 mL
Call For Price
Fisher Chemical M2091 Methyl Ethyl Ketone (Certified ACS) (1 liter) EW-88016-52 Mfr # M2091
COMPARE
78983 1000 Methyl Ethyl Ketone (Certified ACS) (1 liter)
Call For Price
Fisher Chemical M2094 Methyl Ethyl Ketone (Certified ACS) (4 liter) EW-88016-54 Mfr # M2094
COMPARE
78983 4000 Methyl Ethyl Ketone (Certified ACS) (4 liter)
Call For Price
Fisher Chemical M2081 Methyl Ethyl Ketone (Technical) (1 liter) EW-88016-56 Mfr # M2081
COMPARE
78-93-3 1000 Methyl Ethyl Ketone (Technical) (1 liter)
Call For Price
Fisher Chemical M2084 Methyl Ethyl Ketone (Technical) (4 liter) EW-88016-58 Mfr # M2084
COMPARE
78-93-3 4000 Methyl Ethyl Ketone (Technical) (4 liter)
Call For Price
Fisher Chemical M20820 Methyl Ethyl Ketone (Technical) (20 liter) EW-88075-95 Mfr # M20820
COMPARE
78-93-3 20000 Methyl Ethyl Ketone (Technical) (20 liter)
Call For Price
Fisher Chemical M20920 Methyl Ethyl Ketone (Certified ACS) (20 liter) EW-88075-96 Mfr # M20920
COMPARE
78-98-3 20000 Methyl Ethyl Ketone (Certified ACS) (20 liter)
Call For Price
Fisher Chemical M209200 Methyl Ethyl Ketone (Certified ACS) (200 liter) EW-88075-97 Mfr # M209200
COMPARE
78-98-3 200000 Methyl Ethyl Ketone (Certified ACS) (200 liter)
Call For Price
Fisher Chemical M209S4 Methyl Ethyl Ketone (Certified ACS) (4 liter) safetin EW-88076-12 Mfr # M209S4
COMPARE
78-98-3 4000 Methyl Ethyl Ketone (Certified ACS) (4 liter) safetin
Call For Price
COMPARE
78-93-3 1000 Methyl Ethyl Ketone Technical, 1 L
Call For Price
Showing 1- 10 of 10

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat