Meiji Microscope Bodies
Get crisp, high-resolution images with excellent stereoscopic viewing. The 45° inclined head has adjustable diopter and interpupillary controls for easy observing. Rotate body a full 360° and lock into any position. The eyepiece diameters on all models are 25 mm ID x 34 mm OD.
$813.50 - $1,526.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Magnification Description
Availability Pricing
Meiji Techno EMZ-5 Stereozoom microscope bodies; objectives, 0.7x to 4.5x EW-48402-20 Mfr # EMZ-5
COMPARE
Binocular 0.7 to 4.5x Stereozoom microscope bodies; objectives, 0.7x to 4.5x
$1,526.00USD / Each
Meiji Techno EMT-1 Dual-power microscope bodies; objectives, 1x/2x EW-48403-20 Mfr # EMT-1
COMPARE
Binocular 1x/2x Dual-power microscope bodies; objectives, 1x/2x
$813.50USD / Each
Meiji Techno EMT-2 Dual-power microscope bodies; objectives, 1x/3x EW-48403-30 Mfr # EMT-2
COMPARE
Binocular 1x/3x Dual-power microscope bodies; objectives, 1x/3x
$906.00USD / Each