Megger MOM2 Precision Microhmmeters
Available for sale only in the United States Precision low resistance measurements in a rugged, compact meter
  • Up to 220 A
  • Safe testing via DualGround™ method
  • Auto ranging from 1 µ to 1000 m
  • Complies with IEEE and IEC standards
  • Store up to 190 test values and transfer test data to a PC via Bluetooth®
  • Lightweight, rugged design for use out in the field
  • Conduct a full day of testing without battery recharge
This microhmmeter is designed to measure low resistance values with high accuracy in circuit breaker contacts, bus-bar joints, high-current links and more. Designed with safety, ease-of-use and versatility in mind, this unit allows you to make measurements according to the DualGround™ method. This method defines the test object to be grounded on both sides throughout the test giving a safer, faster, and easier workflow.
The easy-to-read LED display provides a combination of analogue arc and dual digital readouts for indication of capacitor charge, and main measurement results. Unit makes the transfer of measurement data simple to your PC via Bluetooth technology for documentation and further analysis.
Applications
Testing circuit breaker contacts
esting connections to the breaker
Testing of bus-bar joints and connections
Low resistance, high current connections:
- Switches
- Disconnecting devices
- Safety ground connections
- Welding points
- Fuses and cables
$9,503.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
AC Current Resolution (mA) DC Voltage Range (Voltage)
Availability Pricing
Megger MOM2 with clamps 200A Microhmmeter w/ 4 and 10 Ft Test Cables EW-20041-38 Mfr # BD-59092
COMPARE
$9,503.00USD / Each