Megger MIT 400/2 Series Insulation Testers
Ideal for electrical and industrial testing
  • Single range, faster continuity testing from 0.01 Ω to 1 MΩ
  • Stabilized insulation test voltage is accurate to +2% -0%
  • Live circuit detection for added protection
The Megger MIT400/2 series insulation and continuity testers offer wide range of applications from electrical installations, cable testing, motor testing, automotive, ESD, panel building, avionics, and maintenance. These compact units provide insulation testing up to 1000 V and 200 GΩ range. A variable range allows any intermediate voltage from 10 to 1000 V in 1 V steps.
Each tester is protected by a rubber molded outer casing and comes with a IP54 weatherproof rating.
$992.50 - $1,386.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Megger MIT400/2 Insulation Tester, CAT IV EW-20026-31 Mfr # 1004-731
COMPARE
Insulation Tester, CAT IV
$992.50USD / Each
Megger MIT410/2 Insulation Tester, CAT IV, PI and DAR EW-20026-32 Mfr # 1004-735
COMPARE
Insulation Tester, CAT IV, PI and DAR
$1,155.00USD / Each
Megger MIT420/2 Insulation Tester, CAT IV, PI and DAR, Data Logging EW-20026-33 Mfr # 1004-739
COMPARE
Insulation Tester, CAT IV, PI and DAR, Data Logging
$1,213.00USD / Each
Megger MIT430/2 Insulation Tester, CAT IV, PI and DAR, Data Logging, Wireless EW-20026-34 Mfr # 1004-740
COMPARE
Insulation Tester, CAT IV, PI and DAR, Data Logging, Wireless
$1,386.00USD / Each