This product is now discontinued, View Related Products
Megger Analog 5kV Insulation Resistance Testers
Get quick, accurate reading of insulation resistance
  • Resilient mounted analogue scale for robustness
  • Voltage range to 600 V indicates auto discharge
  • Measurements can be conducted in a.c. or d.c.
  • Pass/fail overlays for rapid testing
  • Single scale for insulation values avoids operator error
These compact 5kV insulation testers are simple to use, providing you a quick and accurate reading of insulation resistance. The analog display features a maximum reading of 20 GΩ. Four test voltages are available at 500 V, 1 kV, 2.5 kV and 5 kV. The testers are supplied with optional locking button for long term testing.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
AC Voltage Accuracy Max Resistance (G ohms) Description
Availability Pricing
Megger 210962 Model , Set of 3 Test leads, each 10' long leads EW-20010-35 Mfr # 210962
COMPARE
Model , Set of 3 Test leads, each 10' long leads
Call For Price
Megger 218650 BM11 Insulation Resistance Tester EW-20022-35 Mfr # 218650
COMPARE
BM11 Insulation Resistance Tester
Call For Price
Megger MJ15 Analog 5kV Insulation Tester, Hand-Crank Generator/Batteries EW-20022-36 Mfr # 6410-920
COMPARE
±5% of nominal test voltages on 20 MΩ load 20 Analog Insulation Tester Hand Crank Rechargable Battery, 1k Ohm/20 g Ohm
Call For Price
Megger BM15 Analog 5kV Insulation Tester, Battery Operated EW-20022-37 Mfr # 6410-919
COMPARE
±5% of nominal test voltages on 20 MΩ load 20 Compact Analog 5 kV Insulation Tester