Masterflex® Turbine Flowmeter Rate Sensors
Measures low liquid flow rates with high accuracy
  • Low cost plus high accuracy with ±3% of reading
  • Lightweight plastic design enables mounting in any position
These hall effect turbine flow rate sensors are ideal for OEM applications involving low flow liquid monitoring. The low cost coupled with 1/2% repeatability makes it an ideal candidate for replacing dispensing timer systems. Unlike existing timing systems, turbine technology is not influenced by changes in system pressure caused by aging filters. The sensor's standard power and output specifications make it easy to retrofit to existing controllers.
$207.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Min Flow Rate (GPM) Description
Availability Pricing
COMPARE
2 0.13 FT-110 Turboflow Sensor, 0.13 to 2 GPM with 3 Foot Cable, 76-1150 Hz
$207.00USD / Each
COMPARE
1.3 0.13 FT-110 Turboflow Sensor, 0.13-1.3 GPM with Spade Terminal, 58-575 Hz
$207.00USD / Each