Masterflex® Transfer Tubing, Platinum-Cured Silicone
No need to worry about the heat - use for applications up to 460°F
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Pharmaceutical, biotechnology, aerospace, food and beverage, and general lab applications.
Characteristics: Platinum-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent.
Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI; exceeds 3A Sanitary Standards. Tubing is manufactured according to GMP.
Temperature range: -58 to 446°F (-50 to 230°C)
Sterilize: Autoclave or gamma irradiation.
$49.50 - $462.00USD / Pkg of 1
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
15 1/16 1/8
$49.50USD / Pkg of 1
COMPARE
14 3/32 5/32
$82.00USD / Pkg of 1
COMPARE
10 1/8 3/16
$83.50USD / Pkg of 1
COMPARE
21 1/8 1/4
$81.00USD / Pkg of 1
COMPARE
25 1/8 3/8
$183.00USD / Pkg of 1
COMPARE
10 5/32 7/32
$92.50USD / Pkg of 1
COMPARE
9 3/16 1/4
$106.00USD / Pkg of 1
COMPARE
13 3/16 5/16
$95.50USD / Pkg of 1
COMPARE
22 3/16 7/16
$210.00USD / Pkg of 1
COMPARE
17 1/4 1/2
$223.00USD / Pkg of 1
COMPARE
10 5/16 7/16
$174.00USD / Pkg of 1
COMPARE
16 5/16 1/2
$198.00USD / Pkg of 1
COMPARE
20 5/16 9/16
$241.00USD / Pkg of 1
COMPARE
12 3/8 9/16
$216.00USD / Pkg of 1
COMPARE
14 3/8 5/8
$268.00USD / Pkg of 1
COMPARE
10 1/2 11/16
$257.00USD / Pkg of 1
COMPARE
12 1/2 3/4
$329.00USD / Pkg of 1
COMPARE
12 5/8 7/8
$383.00USD / Pkg of 1
COMPARE
9 3/4 1
$462.00USD / Pkg of 1
COMPARE
21 1/8 1/4
$131.00USD / Pkg of 1
Showing 1 - 20 of 28