Masterflex® Transfer Tubing, Peroxide-Cured Silicone
No need to worry about the heat - use for applications up to 460°F
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Applications: Pharmaceutical, biotechnology, food and beverage, aerospace, and general lab applications.
Characteristics: Peroxide-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent.
Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements. Tubing is manufactured according to GMP.
Temperature range: -60 to 460°F (-51 to 238°C)
Sterilize: Autoclave or gamma irradiation.
$40.50 - $354.00USD / Pkg of 1
27 variations of this product are available.
See All Product Options Below
27 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
COMPARE
13 3/16 5/16
$84.00USD / Pkg of 1
COMPARE
10 3/8 1/2
$107.00USD / Pkg of 1
COMPARE
10 1/2 11/16
$196.00USD / Pkg of 1
COMPARE
12 1/2 3/4
$237.00USD / Pkg of 1
COMPARE
12 5/8 7/8
$286.00USD / Pkg of 1
COMPARE
30 1/50 1/12
$59.00USD / Pkg of 1
COMPARE
31 1/32 3/32
$55.00USD / Pkg of 1
COMPARE
15 1/16 1/8
$40.50USD / Pkg of 1
COMPARE
14 3/32 5/32
$50.50USD / Pkg of 1
COMPARE
10 1/8 3/16
$65.00USD / Pkg of 1
COMPARE
10 5/32 5/32
$65.00USD / Pkg of 1
COMPARE
9 3/16 1/4
$73.50USD / Pkg of 1
COMPARE
22 3/16 7/16
$182.00USD / Pkg of 1
COMPARE
8 1/4 5/16
$94.50USD / Pkg of 1
COMPARE
12 1/4 3/8
$93.00USD / Pkg of 1
COMPARE
17 1/4 1/2
$170.00USD / Pkg of 1
COMPARE
10 5/16 7/16
$105.00USD / Pkg of 1
COMPARE
8 1/2 5/8
$165.00USD / Pkg of 1
COMPARE
10 5/8 13/16
$257.00USD / Pkg of 1
COMPARE
9 3/4 1
$354.00USD / Pkg of 1
Showing 1 - 20 of 27