This product is now discontinued, View Related Products
Masterflex® Single-Use Sampling Bottle Cap Assembly
Safely use your own bottles to transfer and store fluids in your work area
  • Create a closed system to collect fluids from your application
  • Create a system with ventilation or to prevent spills
Single use cap assembly for connecting your bottles to your application and collect, transfer and store fluids. Cap assemblies come with Puri-Flex™ biopharma tube/linear assembly with 1/8" ID x 1/4" OD tubing and HDPE cap to fit a variety of bottles. Purchase bottle separately.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material Description
Availability Pricing
COMPARE
38 mm HDPE Masterflex® Single-Use Sterile Sampling Bottle Assembly, PETG Bottle, ; 2 L
Call For Price
COMPARE
53 mm HDPE Masterflex® Single-Use Sterile Sampling Bottle Assembly, PETG Bottle, ; 2 L