Masterflex Replacement Pump Heads for Quattroflow Pump Systems
Keep your Quattroflow™ pump system up and running with these quality replacement pump heads
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Single-use pump heads are constructed with a disposable polypropylene chamber
  • Reusable pump heads are constructed of durable 316L stainless steel
Keep your Quattroflow pump system up and running with these quality replacement pump heads. The quaternary diaphragm design enables risk-free dry-running, self-priming, and a high turn-down ratio. These pump heads can safely and reliably pump plasma products, proteins, monoclonal antibodies, vaccines, and more.
Order the Masterflex® Single-use Quattroflow™ Pump Systems separately.
$545.25 - $3,087.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Disposable Pump Chamber, PP machined for ABS and stainless steel single-use Quattroflow pump systems
$545.25USD / Each
COMPARE
Disposable Pump Chamber; PP machined for stainless steel single-use Quattroflow pump systems
$1,233.50USD / Each
COMPARE
Multiple-Use Pump Chamber; 316L machined for ABS and stainless steel multiuse Quattroflow pump systems
$1,592.50USD / Each
COMPARE
Mutiple-Use Pump Chamber; 316L machined stainless steel multiuse Quattroflow pump systems
$3,087.00USD / Each