Masterflex® Paddle-Wheel In-Line Flowmeters
Accurate measurement with virtually zero pressure drop
  • Paddle wheel technology is accurate, affordable, and relatively easy to install and maintain
  • Ideally suited to open channels and partially filled pipes containing water
  • Factory calibrated for GPM or LPM
These economical in-line flowmeters are battery powered (two AAA batteries included) and thus perfect for remote locations. Enclosure is rated NEMA 4X and can be used in outdoor locations. Sensor is constructed of corrosion-resistant PVDF; in-line NPT(M) fittings are of PP.
$437.00 - $790.00USD / Each
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Min Flow Rate (GPM) Process Connection (NPT(M))
Availability Pricing
COMPARE
4 0.4 0
$440.00USD / Each
COMPARE
8 0.8 0
$453.00USD / Each
COMPARE
30 3
$437.00USD / Each
COMPARE
50 5
$440.00USD / Each
COMPARE
60 6
$526.00USD / Each
COMPARE
40 4
$554.00USD / Each
COMPARE
8 0.8 0
$659.00USD / Each
COMPARE
30 3
$679.00USD / Each
COMPARE
50 5
$659.00USD / Each
COMPARE
2.6 0.26 0
$449.00USD / Each
COMPARE
8 0.8 0
$453.00USD / Each
COMPARE
18.5 1.8
$440.00USD / Each
COMPARE
29 3
$453.00USD / Each
COMPARE
53 5.3
$440.00USD / Each
COMPARE
66 6.6
$516.00USD / Each
COMPARE
40 4
$511.00USD / Each
COMPARE
40 4
$538.00USD / Each
COMPARE
29 3
$694.00USD / Each
COMPARE
66 6.6
$773.00USD / Each
COMPARE
79 8
$790.00USD / Each
Showing 1 - 20 of 28