This product is now discontinued, View Related Products
Manifold Mixing Solenoid Valves
Stand up to aggressive fluids
  • Excellent for multiple liquid or gas control systems with aggressive or high-purity fluids
  • PTFE or PEEK wetted parts
  • Continuous duty and high cycle life
  • Suitable for pressure or vacuum service
These normally closed solenoid mixing valves feature low power consumption, minimal dead volume, and fast response time for all your mixing needs. All valves come with 15" (26 gauge) PTFE coated lead wires. Special orders are available for valves with different diaphragm and body material, or larger flow values — call for more information.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Power (VDC) Wetted Materials
Availability Pricing
Manifold Mixing Solenoid Valve, PTFE; 3 In/1 Out; 24VDC EW-01356-06 Mfr # 080T324-62
COMPARE
3 Solenoid, 3 inlet, 1 outlet 24 PTFE