Masterflex® Magnetic Drive Centrifugal Pump

Heavy-duty impeller spindle and fluted impeller combine for efficient cooling
  • Pumps available with or without 56 C-face motors — install the motor you need!
  • Pumps with motor are close-coupled for easy installation and maintenance
  • Ideal for fluid transfer or circulation — use pumps for chemical processing plants and water and wastewater treatment centers
  • Encapsulated magnet
All pumps with motors are set up for hardwiring and are rated for 50/60 Hz duty.
$1,069.00 - $1,781.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Description
Availability Pricing
COMPARE
32 37 Mag Drive PP Centrifugal Pump Head; 32 GPM/11.3 ft
$1,069.00USD / Each
COMPARE
32 37 Mag Drive PP Centrifugal Pump; 32 GPM/11.3 ft, 115/230V
$1,474.00USD / Each
COMPARE
32 37 Mag Drive PP Centrifugal Pump; 32 GPM/11.3 ft, 230/460V
$1,391.00USD / Each
COMPARE
68 59 Mag Drive PP Centrifugal Pump; 68 GPM/59 ft, 115/230V
$1,781.00USD / Each
COMPARE
68 59 Mag Drive PP Centrifugal Pump; 68 GPM/59 ft, 230/460V
$1,648.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat