Masterflex® L/S® Easy-Load® II Pump Heads for Precision Tubing
Automatic retention holds tubing securely in place with no manual adjustment—just load the tubing and start the pump
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Four-roller rotor improves pressure performance and mechanical stability while reducing pulsation
  • Model available with integrated open-head sensor–connects directly to sensor-equipped drives
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S® Precision pump tubing sizes
These pump heads accept the six L/S® precision tubing sizes. For stacking multiple heads (up to the specified limits of your drive) order two- three- or four-channel hardware separately under "Accessories". When two Easy-Load® II pump heads are stacked rollers are offset by 45°--combine the outlets from two stacked heads for a 95% reduction in flow pulsation.
$398.00 - $596.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Occlusion Open-Head Sensor Rotor Material
Availability Pricing
COMPARE
Fixed Yes Stainless Steel
$596.00USD / Each
COMPARE
Fixed No Cold-rolled Steel
$398.00USD / Each
COMPARE
Fixed No Stainless Steel
$504.00USD / Each
COMPARE
Adjustable No Stainless Steel
$542.00USD / Each