Masterflex® Hose Clamps, Nylon
Secure tubing to connectors, pumps, glassware, and filtration devices
  • Use to fasten pressure, vacuum, and liquid tubing lines
  • Nylon is corrosion-proof, chemical resistant, and non-conductive
  • Works with all types of plastic and rubber tubing
  • Always in stock--most formulations and sizes available to ship same day
Reusable clamp squeezes shut for a leakproof seal—press the sides to release the clamp. Order the size that suits your needs, or the handy kit to handle multiple projects.
$33.00 - $56.00USD / Pkg of 100
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
For Tubing OD (in) Max Tubing OD (in)
Availability Pricing
COMPARE
1/4 19/64
$33.00USD / Pkg of 100
COMPARE
23/64 13/32
$33.00USD / Pkg of 100
COMPARE
13/32 15/32
$40.75USD / Pkg of 100
COMPARE
15/32 17/32
$38.25USD / Pkg of 100
COMPARE
17/32 39/64
$42.75USD / Pkg of 100
COMPARE
19/32 43/64
$41.55USD / Pkg of 100
COMPARE
43/64 25/32
$38.00USD / Pkg of 100
COMPARE
47/64 53/64
$49.25USD / Pkg of 100
COMPARE
7/8 63/64
$38.00USD / Pkg of 100
COMPARE
61/64 1
$56.00USD / Pkg of 100
COMPARE
1 1
$56.00USD / Pkg of 100
COMPARE
1/4 1 39/64
$56.00USD / Pkg of 65
Showing 1- 12 of 12