Masterflex® Hose Clamps, Acetal Copolymer
Reusable white acetal copolymer clamps will not rust or corrode
  • Resists corrosion
  • Always in stock--most formulations and sizes available to shiop same day
Recommended for continuous use from ambient temperature to 200ºF (93ºC). Clamps are nonconducting and stable in a wide variety of chemicals.
Once set, clamps stay put. To use, just position clamp on hose or tubing bundle and press the jaws together with a pair of pliers.
$33.25 - $103.00USD / Pkg of 50
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Maximum ID (in) Minimum ID (in)
Availability Pricing
COMPARE
0.256 0.228
$41.00USD / Pkg of 100
COMPARE
0.394 0.351
$41.25USD / Pkg of 100
COMPARE
0.342 0.307
$40.75USD / Pkg of 100
COMPARE
0.452 0.404
$40.75USD / Pkg of 100
COMPARE
0.511 0.449
$40.75USD / Pkg of 100
COMPARE
0.586 0.513
$41.00USD / Pkg of 100
COMPARE
0.65 0.569
$40.75USD / Pkg of 100
COMPARE
0.707 0.616
$46.75USD / Pkg of 100
COMPARE
0.801 0.702
$46.75USD / Pkg of 100
COMPARE
0.848 0.759
$46.75USD / Pkg of 100
COMPARE
0.899 0.779
$46.75USD / Pkg of 100
COMPARE
0.989 0.859
$46.75USD / Pkg of 100
COMPARE
1.072 0.928
$57.00USD / Pkg of 100
COMPARE
1.135 0.985
$56.50USD / Pkg of 100
COMPARE
1.296 1.123
$33.25USD / Pkg of 50
COMPARE
1.405 1.218
$40.75USD / Pkg of 50
COMPARE
1.405 0.228
$103.00USD / Each
COMPARE
0.256 0.256
$46.75USD / Pkg of 100
COMPARE
0.291 0.269
$46.00USD / Pkg of 100
COMPARE
0.44 0.387
$46.75USD / Pkg of 100
Showing 1- 20 of 20