Masterflex® Correlated Variable-Area Flowmeters with Valves, High-Flow, 65-mm
Don’t sacrifice suitability for precision
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
$145.00 - $315.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fluid Type Material Housing Max Flow Rate (LPM)
Availability Pricing
COMPARE
Air Aluminum 23.169
$145.00USD / Each
COMPARE
Air Aluminum 42.094
$145.00USD / Each
COMPARE
Air Aluminum 58.5
$145.00USD / Each
COMPARE
Water PTFE 0.522
$315.00USD / Each
COMPARE
Water PTFE 0.798
$315.00USD / Each
COMPARE
Air 316 SS 23.169
$282.90USD / Each
COMPARE
Air 316 SS 42.094
$282.90USD / Each
COMPARE
Air 316 SS 58.5
$282.90USD / Each
COMPARE
Air Brass 42.094
$186.40USD / Each
COMPARE
Air Brass 58.5
$186.40USD / Each
Showing 1- 10 of 10