Masterflex® Correlated Variable-Area Flowmeter with High and Low Optical Alarms
Protect equipment with automatic flow rate monitoring via high and low alarms
  • Protect your equipment from damage caused by extreme flow rates
  • Features dual-mounted LED receiver/sensors with optical sensors
These 150-mm valved flowmeters feature two noninvasive optical sensors that easily glide up and down both sides of the meter to set high and low flow rates. The meter is ideal for use with an alarm, valve, or other devices to control flow with use of NPN and PNP transistor outputs. The compact design and ease of operation make this meter perfect for laboratory and industrial uses.
$706.25 - $855.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Material Housing Max Flow Rate (mL/min) Description
Availability Pricing
COMPARE
Aluminum 264 Variable-Area Flowmeter, Correlated Reading with Dual Optical Alarms, Aluminum Housing, 264 mL/min Air
$706.25USD / Each
COMPARE
Aluminum 1682 Variable-Area Flowmeter, Correlated Reading with Dual Optical Alarms, Aluminum Housing, 1,682 mL/min Air
$706.25USD / Each
COMPARE
Aluminum 2313 Variable-Area Flowmeter, Correlated Reading with Dual Optical Alarms, Aluminum Housing, 2,313 mL/min Air
$706.25USD / Each
COMPARE
Aluminum 4662 Variable-Area Flowmeter, Correlated Reading with Dual Optical Alarms, Aluminum Housing, 4,662 mL/min Air
$706.25USD / Each
COMPARE
Stainless Steel 814 Variable-Area Flowmeter, Correlated Reading with Dual Optical Alarms, Stainless Steel Housing, 814 mL/min Air
$842.75USD / Each
COMPARE
Stainless Steel 2313 Variable-Area Flowmeter, Correlated Reading with Dual Optical Alarms, Stainless Steel Housing, 2,313 mL/min Air
$842.75USD / Each
COMPARE
Stainless Steel 4662 Variable-Area Flowmeter, Correlated Reading with Dual Optical Alarms, Stainless Steel Housing, 4,662 mL/min Air
$842.75USD / Each
COMPARE
Stainless Steel 16737 Variable-Area Flowmeter, Correlated Reading with Dual Optical Alarms, Stainless Steel Housing, 16,737 mL/min Air
$842.75USD / Each
COMPARE
Stainless Steel 45227 Variable-Area Flowmeter, Correlated Reading with Dual Optical Alarms, Stainless Steel Housing, 45,227 mL/min Air
$855.00USD / Each
Showing 1- 9 of 9