Masterflex® Correlated Flowmeters with High-Res Valves, 150-mm Scale
Don’t sacrifice suitability for precision
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).
$270.80 - $469.00USD / Each
61 variations of this product are available.
See All Product Options Below
61 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Material Housing Max Flow Rate (mL/min)
Availability Pricing
COMPARE
PTFE and Kel-F® PTFE 30
$428.00USD / Each
COMPARE
PTFE and Kel-F® PTFE 825
$428.00USD / Each
COMPARE
PTFE and Kel-F® PTFE 1093
$428.00USD / Each
COMPARE
PTFE and Kel-F® PTFE 11356
$422.00USD / Each
COMPARE
Aluminum Aluminum 18.7
$270.80USD / Each
COMPARE
Aluminum Aluminum 49.1
$270.80USD / Each
COMPARE
Aluminum Aluminum 137
$270.80USD / Each
COMPARE
Aluminum Aluminum 92
$270.80USD / Each
COMPARE
Aluminum Aluminum 374
$270.80USD / Each
COMPARE
Aluminum Aluminum 4562
$270.80USD / Each
COMPARE
Aluminum Aluminum 7825
$270.80USD / Each
COMPARE
316 Stainless Steel 316 Stainless Steel 60.5
$469.00USD / Each
COMPARE
316 Stainless Steel 316 Stainless Steel 264
$469.00USD / Each
COMPARE
316 Stainless Steel 316 Stainless Steel 844
$469.00USD / Each
COMPARE
316 Stainless Steel 316 Stainless Steel 4562
$469.00USD / Each
COMPARE
316 Stainless Steel 316 Stainless Steel 7825
$469.00USD / Each
COMPARE
316 Stainless Steel 316 Stainless Steel 16737
$469.00USD / Each
COMPARE
Chrome-plated brass Brass 60.5
$288.90USD / Each
COMPARE
Chrome-plated brass Brass 92
$281.70USD / Each
COMPARE
Chrome-plated brass Brass 4562
$288.90USD / Each
Showing 1 - 20 of 61