Masterflex® B/T® PerfectPosition® Pump Tubing, BioPharm Platinum-Cured Silicone

Ensure optimal performance from your Masterflex® B/T series pump
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Platinum-cured silicone
  • Ultra-smooth inner surface
  • Fewer extractables
  • Minimizes particle entrapment
  • Perfect for lab, biotechnology, & pharmaceutical applications
  • Long tubing life
  • USP Class VI; Exceeds 3A Standards
  • FDA Compliant
Formulation: Tygon 3350 silicone, platinum cured.
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications.
Characteristics: Ultra-smooth; minimizes bacterial growth. Good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, and nontoxic. Transparent.

NOTE:The below tubing is not designed to work with the B/T Perfect Position Pump Systems.
$4,628.00 - $5,183.00USD / Pkg of 1

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
B/T Masterflex tubing size Hose Barb Size (in) Max Flow Rate (mL/min)
Availability Pricing
COMPARE
90 3/4 42000
$5,183.00USD / Pkg of 1
COMPARE
92 1 45000
$4,628.00USD / Pkg of 1
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat