Marsh Bellofram Type 1000 I/P Transducers
High-flow capacity up to 12 scfm
  • 4 to 20 mA Input
  • Mounts in any angle
  • Small size and lightweight
  • External zero and span adjustments
These transducers are an electro-pneumatic device that reduces supply pressure to a regulated output pressure directly proportional to an electrical input signal. These transducers are also field-reversible by reversing the polarity of the signal leads and recalibrating. After calibration, the transducer will then provide a minimum of it''s full rated output pressure upon input signal failure. These units are typically used in HVAC-R systems, petrochemical processing, and slurry processing.
$480.00 - $605.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Control Range (high) (PSI) Control Range (low) (PSI) Description
Availability Pricing
Marsh Bellofram 961-086-000 Pressure Transmitter, I to P, 1-9VDC-3-27PSI E/P (Volts To Press) EW-68458-21 Mfr # 961-086-000
COMPARE
27 3 Pressure Transmitter, I to P, 1-9VDC-3-27PSI E/P (Volts To Press)
$480.00USD / Each
Marsh Bellofram 961-111-000 Pressure Transmitter, I to P, 4-20MA-3-120PSI I/P (Curr To Press) EW-68458-24 Mfr # 961-111-000
COMPARE
120 3 Pressure Transmitter, I to P, 4-20MA-3-120PSI I/P (Curr To Press)
$605.00USD / Each
Marsh Bellofram 961-117-000 Pressure Transmitter, I to P 4-20MA-2-60PSI I/P (Curr To Press) EW-68458-26 Mfr # 961-117-000
COMPARE
60 2 Pressure Transmitter, I to P 4-20MA-2-60PSI I/P (Curr To Press)
$605.00USD / Each
Marsh Bellofram 961-070-000 Pressure Transmitter, I to P; 3-15 PSI, 4-20 mA
Always in Stock
EW-68826-00 Mfr # 961-070-000
COMPARE
15 3 Pressure Transmitter, I to P; 3-15 PSI, 4-20 mA
$495.00USD / Each
Marsh Bellofram 961-074-000 Pressure Transmitter, I to P;3-27 PSI, 4-20 mA EW-68826-02 Mfr # 961-074-000
COMPARE
27 3 Pressure Transmitter, I to P;3-27 PSI, 4-20 mA
$495.00USD / Each
Marsh Bellofram 961-075-000 Pressure Transmitter, I to P;6-30 PSI, 4-20 mA
Always in Stock
EW-68826-04 Mfr # 961-075-000
COMPARE
30 6 Pressure Transmitter, I to P;6-30 PSI, 4-20 mA
$495.00USD / Each