Mark-10 R52 Series Static Torque Sensors
Rugged aluminum housing
  • Universal mounting
  • Used for torque tool calibration or as a component of a torque testing system
Calibrate torque tools with this torque measurement sensor, or use it as a component of a torque testing system. Tapped holes on every surface allow for easy mounting to a work bench, or fixture within a testing system.
Table Mounting Kit and 3/8" Jacob's chuck available separately.
$910.00 - $975.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (oz-in)
Availability Pricing
Mark-10 MR52-50Z Torque Sensor, 50 ozFin / 3600 gFcm / 36 kgFmm / 350 Nmm / 35 Ncm EW-25302-11 Mfr # MR52-50Z
COMPARE
50
$975.00USD / Each
Mark-10 MR52-12 Torque Sensor, 12 lbFin / 1 lbFft / 192 ozFin / 135 Ncm / 1.35 Nm EW-25302-12 Mfr # MR52-12
COMPARE
192
$910.00USD / Each