M-System Mini-M Plug-in Signal Conditioners
  • Both Mini-M and Mini-MW use a compact-size, plug-in socket base for quick installation or replacement of module without disturbing wiring
  • 2000 VAC Isolation
  • Standard DIN rail or surface mounting
  • Base socket included with modules
  • CE marking and UL/C-UL Class I Div 2 approval
M-System's Mini-M Series Signal Conditioners are designed to accept the largest range of process signal inputs and provide a standard and non-standard DC output.
The Mini-MW Series Signal Splitters provide a second isolated output of independent range, which give you the flexibility to add sophisticated distributed control or MIS monitoring to a local loop, without a worry about mismatched impedance, or the threat of one problem system's output signal impacting the other.
Both Mini-M and Mini-MW use a compact-size, plug-in socket base for quick installation or replacement of module without disturbing wiring.
ITEM(S) ON SALE
$170.21 - $490.87USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Input type Power Supply (VDC)
Availability Pricing
M-System M2SN-1AA/UL 4/20MA in & out 1 channel signal cond EW-68467-31 Mfr # M2SN-1AA/UL
COMPARE
Loop-powered
$170.21USD / Each
$200.25USD / Each
M-System M2SN-2AA 4/20MA in & out 2 channel signal cond EW-68467-32 Mfr # M2SN-2AA/UL
COMPARE
Loop-powered
$190.82USD / Each
$224.50USD / Each
M-System M2VS-AA-R/UL 4/20MA in & out 24VDC isolator EW-68467-33 Mfr # M2VS-AA-R/UL
COMPARE
24
$234.81USD / Each
$276.25USD / Each
M-System M2VS-AA-M2 4/20MA in & out 100-240VAC isolator EW-68467-34 Mfr # M2VS-AA-M2/UL
COMPARE
$226.73USD / Each
$266.75USD / Each
M-System M2RS-4A-R/UL 0-100F Rtd Transmitter Platinum Rtd 100 Ohm 4-20 Ma24V Dc EW-68467-37 Mfr # M2RS-4A-R/UL 0-100F
COMPARE
RTD 24
$232.47USD / Each
$273.50USD / Each
M-System M2MS-A-R Potentiometer Transmitter 0/10kohm (Min. Span 100ohm) 4-20 Ma24V Dc EW-68467-39 Mfr # M2MS-A-R/UL
COMPARE
24
$249.26USD / Each
$293.25USD / Each
M-System M2MS-A-M2 Mini-M Potentiometer Transmitter; 4-20mA/100-240VAC EW-68467-40 Mfr # M2MS-A-M2/UL
COMPARE
$258.40USD / Each
$304.00USD / Each
M-System M2DYS-24A-R Isolated Currnt Loop Supply 24V/22mA 4-20 Ma 4-20 Ma24V Dc EW-68467-43 Mfr # M2DYS-24A-R/UL
COMPARE
24
$215.47USD / Each
$253.50USD / Each
M-System M2DYS-24A-M2 Isolated Curnt Loop Supply 24V/22mA 4-20 Ma 4-20 Ma100 - 240V Ac EW-68467-44 Mfr # M2DYS-24A-M2/UL
COMPARE
$215.47USD / Each
$253.50USD / Each
M-System M2AP-A1-R/UL 0-1KHZ Dc/Frequency Converter 4-20 Ma Open Collector (Max. 1 Khz) EW-68467-46 Mfr # M2AP-A1-R/UL 0-1KHZ
COMPARE
24
$266.26USD / Each
$313.25USD / Each
M-System W2VS-AAA-R Signal Splitter 4-20 Ma 4-20 Ma24V Dc EW-68467-49 Mfr # W2VS-AAA-R/UL
COMPARE
24
$317.47USD / Each
$373.50USD / Each
M-System W2MS-AA-R Potentiometer Transmitter; Dual-Output/4-20mA/100-240VAC EW-68467-52 Mfr # W2MS-AA-R/UL
COMPARE
24
$339.57USD / Each
$399.50USD / Each
M-System M-SYSTEM MXLCF SIGNAL COND EW-68469-44 Mfr # MXLCF-S2Z1-M2
COMPARE
$490.87USD / Each
$577.50USD / Each
Showing 1- 13 of 13