M-System Low Cost Signal Conditioners
M-System's M5 / B5 / W5 Series signal conditioners are designed to accept a wide variety of process signal input and provide a standard and non-standard DC output. Only 1.6'' (41mm) deep modules can be installed anywhere, even behind the panel cover.
Th M5 series features signal channel four-wire models, while the B5 series are two-wire models. Model B5SN input loop powered isolator can tap into the existing 4-20mA loops without needing to add additional wiring for power. Model B5VS 2-wire output loop powered isolator is designed to accept a DC voltage or current signal input and provide a 4-20mA output. The W5 series signal splittes provide two isolated DC outputs of independent ranges.
ITEM(S) ON SALE
$131.32 - $224.82USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Input type
Availability Pricing
M-System W5DY-AA-R Current Loop Signal Splitter, 4-20 mA EW-25751-12 Mfr # W5DY-AA-R
COMPARE
$198.47USD / Each
$233.50USD / Each
M-System M5VS-AA-R Isolator, 4-20mA, 24V DC EW-68467-54 Mfr # M5VS-AA-R
COMPARE
$133.23USD / Each
$156.75USD / Each
M-System M5VS-44-R Isolator, 0-10V DC, 24V DC EW-68467-55 Mfr # M5VS-44-R
COMPARE
$134.30USD / Each
$158.00USD / Each
M-System M5VS-4A-R M5VS-4A-4 Isolator, 0-10V DC, 24V DC EW-68467-56 Mfr # M5VS-4A-R
COMPARE
$133.02USD / Each
$156.50USD / Each
M-System W5RS-4AA-R 0-250F RTD Signal Splitter 4-20mA, 24V DC EW-68467-58 Mfr # W5RS-4AA-R 0-250F
COMPARE
RTD
$224.82USD / Each
$264.50USD / Each
M-System W5MS-AA-4 Potentiomter Signal Splitter 4-20mA, 24V DC EW-68467-59 Mfr # W5MS-AA-R
COMPARE
$224.82USD / Each
$264.50USD / Each
M-System B5VS-A Two-Wire Isolator, 4-20mA EW-68467-60 Mfr # B5VS-A
COMPARE
$131.32USD / Each
$154.50USD / Each
M-System B5MS Two-Wire Transmitter, 100-10k ohm potentiometer in, 4-20 mA out EW-68467-63 Mfr # B5MS
COMPARE
100Ω to 10kΩ Potentiometer
$149.17USD / Each
$175.50USD / Each
M-System M5RS-4A-R 0-100C RTD Transmitter, 4-20mA, 24V EW-90440-76 Mfr # M5RS-4A-R 0-100C
COMPARE
RTD
$157.03USD / Each
$184.75USD / Each
M-System M5DY-A-R Isolated Current Loop Supply, 24 V/22mA, 24V DC EW-94470-21 Mfr # M5DY-A-R
COMPARE
$144.71USD / Each
$170.25USD / Each
M-System M5SN-AA Loop Powered Isolator, 4-20mA EW-94710-32 Mfr # M5SN-AA
COMPARE
$161.92USD / Each
$190.50USD / Each
Showing 1- 11 of 11