M-System Lightning Surge Protectors
Don’t let lightning take systems down
  • Absorb only the lightning surges with no interruption of the instrumentation signal
  • Lightweight, easy-to-handle, plug-in construction
  • Superior protection and selection across a wide range of applications
  • Remove and test without disconnecting wires
  • Base socket connects input-output signals when an element is removed
ITEM(S) ON SALE
$96.26 - $118.36USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
M-System MDP-24T Lightning/Surge Protector; 4-20mA, Pulse, Photovoltaic EW-43002-02 Mfr # MDP-24T
COMPARE
$118.36USD / Each
$139.25USD / Each
M-System MD7ST-24FF10 Ultra-slim lightning & surge protector for 4-20 mA and pulse signal EW-68467-62 Mfr # MD7ST-24FF10
COMPARE
$116.45USD / Each
$137.00USD / Each
M-System MDP-24-1 Lightning & Surge Protector For 4-20mA And Pulse Signal EW-68467-67 Mfr # MDP-24-1/UL
COMPARE
$96.26USD / Each
$113.25USD / Each
M-System MDP-100 Lightning & Surge Protector For Ac/Dc Power Line Up To 1A EW-68467-68 Mfr # MDP-100
COMPARE
$96.47USD / Each
$113.50USD / Each
M-System MAKF-120 One-Port Lightning/Surge Protector; Power Supply/120VAC EW-68467-69 Mfr # MAKF-120
COMPARE
$107.95USD / Each
$127.00USD / Each