Lovibond MD100 Single Parameter Colorimeters
Uses popular Powder Pack™, tablet, and liquid chemistries
  • Large backlit display for low light conditions
  • Sixteen-point automatic data storage with time and date stamp
  • Ergonomic, waterproof design for easy hand-held use in lab, field, or plant
The Lovibond MD100 series colorimeters are available in a wide variety of parameters using powder-pack chemistries. These rugged, waterproof meters feature a large backlit display, real-time clock, data logging capabilities, and a user calibration mode which allows you to calibrate against prepared standards.
(Note: Lovibond is the new name for Orbeco-Hellige--exactly the same product with a new name.)
$575.00 - $590.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Lovibond MD100 Colorimeter, Chlorine/PH Kit EW-99561-52 Mfr # 278030
COMPARE
Colorimeter, Chlorine/PH Kit
$575.00USD / Each
Lovibond MD100 Colorimeter Manganese LR Kit EW-99561-88 Mfr # 276105
COMPARE
Colorimeter Manganese LR Kit
$590.00USD / Each