Lightnin LB2 LabMaster Mixer Kits
Take the guesswork out of mixing
  • Digitally controlled motor provides high torque over a wide speed range
  • Constant speed maintained by continuous feedback to the motor
  • Smart logic transmission control automatically adjusts speed to optimize mixing
These laboratory and industrial mixers feature a maintenance-free high-efficiency brushless DC motor with a controller designed for safety. If torque demand spikes, the controller will automatically reduce the speed or shut off the motor to prevent overload or overheat conditions. Integrated overload protection prevents damage to electronics, motor, and transmission in the event of an overload or overheat situation. Mixer powers up in "pause mode", ramps up to speed to avoid splashing, and remains off after power interruption for added operational safety.
Choose a kit with the mounting option that works best with your application. The rod mount option includes a support rod and attachment hardware, perfect if you already have a clamp and support stand setup. The universal clamp option includes a support rod with hardware and a clamp which is compatible with both support stands and tank lips. The mechanical seal option mounts to a drum or vessel with a 3" or 4" sanitary flange.
$4,915.00 - $7,095.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Lightnin LabMaster LB2 Digital Laboratory Mixer Kit, Universal Mount; 115 VAC, 50/60 Hz EW-50800-02 Mfr # 873799PSP
COMPARE
Digital Laboratory Mixer Kit, Universal Mount; 115 VAC, 50/60 Hz
$5,670.00USD / Each
Lightnin LabMaster LB2 Digital Laboratory Mixer Kit, Universal Mount; 230 VAC, 50/60 Hz EW-50800-04 Mfr # 873800PSP
COMPARE
Digital Laboratory Mixer Kit, Universal Mount; 230 VAC, 50/60 Hz
$5,965.00USD / Each
Lightnin LabMaster LB2 Digital Laboratory Mixer Kit, Rod Mount; 115 VAC, 50/60 Hz EW-50800-06 Mfr # 873801PSP
COMPARE
Digital Laboratory Mixer Kit, Rod Mount; 115 VAC, 50/60 Hz
$4,915.00USD / Each
Lightnin LabMaster LB2 Digital Laboratory Mixer Kit, Mechanical Seal; 115 VAC, 50/60 Hz EW-50800-22 Mfr # 873817PSP
COMPARE
Digital Laboratory Mixer Kit, Mechanical Seal; 115 VAC, 50/60 Hz
$7,095.00USD / Each
Lightnin LabMaster LB2 Digital Laboratory Mixer Kit, Mechanical Seal; 230 VAC, 50/60 Hz EW-50800-24 Mfr # 873818PSP
COMPARE
Digital Laboratory Mixer Kit, Mechanical Seal; 230 VAC, 50/60 Hz
$7,095.00USD / Each