Lightnin GP2 Laboratory Digital Mixers
A built-in low energy LED light let you easily view the mixing sample
  • Ideal for simple blending, solid suspension, high-viscosity, and gas-liquid applications
  • Integrated digital control and feedback system provide precise control of the mixing process Through-shaft design allows for easy shaft height adjustment
Designed for multifunctional open tank applications, these Lightnin GP2 mixers have speeds up to 2000 rpm and 144 in-oz maximum torque. Digital control and feedback system coupled with Lighnin impellers produce consistent results that is critical for validation and scale-up operations.
Through-shaft design accepts 3/8” (10 mm) dia shaft and offers quick shaft length adjustments, resulting in less downtime. Operate mixers in clockwise or counterclockwise direction or in agitate cycle along with a timer that stops the mixer when time is complete. Mixers feature brushless DC motor, automatic overload protection, and overtemperature protection to ensure low maintenance and long service life. Chemical-resistant, IP42-rated, keypad makes cleanup simple.
$3,210.00 - $3,945.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Lightnin Labmaster GP2 Digital Laboratory Mixer Kit, Universal Mount; 115/230 VAC, 50/60Hz EW-50800-12 Mfr # 873856PSP
COMPARE
Digital Laboratory Mixer Kit, Universal Mount; 115/230 VAC, 50/60Hz
$3,945.00USD / Each
Lightnin Labmaster GP2 Digital Laboratory Mixer Kit, Universal Mount; Power Cable Kit EW-50800-14 Mfr # 873857PSP
COMPARE
Digital Laboratory Mixer Kit, Universal Mount; Power Cable Kit
$3,945.00USD / Each
Lightnin Labmaster GP2 Digital Laboratory Mixer Kit, Rod Mount; 115/230 VAC, 50/60Hz EW-50800-16 Mfr # 873805PSP
COMPARE
Digital Laboratory Mixer Kit, Rod Mount; 115/230 VAC, 50/60Hz
$3,210.00USD / Each