AEMC® Instruments Light Meters with Mountable Probe
Probe attaches to top of meter for optional one-handed operation
  • Easy one-hand operation
  • Designed to measure a wide range of lighting types
  • Removable sensor for remote reading
  • Measures in footcandles (fc) or lux
  • Measures incandescent lighting
  • Cosine corrected
  • Hold function
  • Max function (CA811)
  • Peak function (CA813)
  • CIE photoptic (human eye) response
  • 2000-count backlit LCD
  • Lightweight and compact
  • Removable protective sensor cover
  • Includes rugged, shockproof, protective and dirt resistant gray cover
Light meters feature optical sensors designed to match the response of the human eye—ideal for assessment of employee workspace light levels. Powerful meters feature an ergonomic housing, large display with HOLD option, and intuitive setting controls. Both models include a shockproof protective holster to protect meter from dirt and damage.
$121.60 - $192.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Measurement (Lux)
Availability Pricing
AEMC CA811 Digital Light Meter with Mountable Sensor EW-86533-00 Mfr # 2121.2
COMPARE
20000
$121.60USD / Each
AEMC CA813 High-range light meter with mountable pensor EW-86533-10 Mfr # 2121.21
COMPARE
200000
$192.00USD / Each