Lascar Electronics Compact USB Data Loggers, Process
Direct-to-USB connection and easy-to-use functionality
  • User-programmable alarm thresholds
  • All data loggers come with software
Simply insert any of these compact, 4.5" (11.4 cm) L loggers into your computer USB port, name it, choose your required sampling rate, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
ITEM(S) ON SALE
$86.20USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DC Current Accuracy (ADC) DC Current Resolution (ADC) Max Current (ADC)
Availability Pricing
Lascar Electronics EL-USB-3 USB Voltage Data Logger, Process, 32,000+ readings, 1 year battery life EW-23039-54 Mfr # EL-USB-3
COMPARE
$86.20USD / Each
$107.75USD / Each
Lascar Electronics EL-USB-4 USB Current Data Logger, Process, 32,000+ readings, 1 year battery life EW-23039-56 Mfr # EL-USB-4
COMPARE
±1% of reading 0.02
$86.20USD / Each
$107.75USD / Each