Large Displays for Current, Voltage, Process, Strain Gage, and Temperature

Improve process visability and shop floor productivity
  • LPAX modules feature large 1.5" (38 mm) red display readable from 70 feet (21 meters)
  • EPAC modules feature extra-large 4" (101 mm) red display readable from 180 feet (55 meters)
  • User programmable inputs
  • Field-installable option cards
  • NEMA 4X / IP65-rated front panels
Combining an MPAX input module with a large (LPAX) or extra-large (EPAX) display gives you a display size meter with the same functionality as the analog input PAX meters described on page 787. These large displays make an excellent productivity tool by providing your shop floor with easy to read plant status in real time.

We list below some of the more popular models with analog input, but others are available so check with our applications representative if you have a specific need not addressed. These diplays are field upgradeable with option cards shown below, including communications, setpoints and retransmitted analog outputs to provide flexibility and affordability as your needs grow.
$734.00 - $1,083.50USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Red Lion 68485-72 + 68485-44 5 Digit Display with MPAXP000 Input Module EW-68495-09 Mfr # 68485-72 + 68485-44
COMPARE
5 Digit Display with MPAXP000 Input Module
$734.00USD / Each
Red Lion 68485-73 + 68485-44 5 Digit Display with MPAXP010 Input Module EW-68495-10 Mfr # 68485-73 + 68485-44
COMPARE
5 Digit Display with MPAXP010 Input Module
$774.50USD / Each
Red Lion 68485-72 + 68485-73 5 Digit Display with MPAXP000 Input Module EW-68495-11 Mfr # 68485-72 + 68485-73
COMPARE
5 Digit Display with MPAXP000 Input Module
$1,083.50USD / Each
Red Lion 68485-77 + 68485-44 5 Digit Display with MPAXT000 Input Module EW-68495-12 Mfr # 68485-77 + 68485-44
COMPARE
5 Digit Display with MPAXT000 Input Module
$734.00USD / Each
Red Lion 68485-77 + 68485-73 5 Digit Display with MPAXT000 Input Module EW-68495-14 Mfr # 68485-77 + 68485-73
COMPARE
5 Digit Display with MPAXT000 Input Module
$1,083.50USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat